| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09020682.hg.1 |
ENST00000421699.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09020682.hg.1 |
NM_004486.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09020682.hg.1 |
TC09001628.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000421699.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000450617.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000458730.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000490257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000609374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000611957.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
NM_004486.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000421699.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000450617.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000458730.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000490257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000609374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
ENST00000611957.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
NM_004486.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179577.hg.1 |
TC0900011631.hg.1 |
Clariom D TC |