| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023360.hg.1 |
ENST00000298630.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023360.hg.1 |
ENST00000368622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023360.hg.1 |
ENST00000368625.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023360.hg.1 |
ENST00000478542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023360.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023360.hg.1 |
TC10001771.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012611.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
ENST00000298630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
ENST00000368622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
ENST00000462160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
NM_173575.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000188268.hg.1 |
TC1000012611.hg.1 |
Clariom D TC |