Assay Details
| Gene Symbol: STK32C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011539694.1 | XP_011537996.1 | 2 - 3 | 136 | 95 | |
| XM_011539695.1 | XP_011537997.1 | 2 - 3 | 337 | 95 | ||
| XM_011539696.1 | XP_011537998.1 | 2 - 3 | 144 | 95 | ||
| GenBank mRNA | AK308956.1 | - | 2 - 3 | 475 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011539694.1, XM_011539695.1, XM_011539696.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000298630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000368622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | ENST00000462160.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012611.hg.1 | NM_173575.3 | Refseq |

STK32C
Genomic Map