| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04020220.hg.1 |
ENST00000265148.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04020220.hg.1 |
ENST00000380026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04020220.hg.1 |
ENST00000394771.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04020220.hg.1 |
NM_001813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04020220.hg.1 |
TC04001436.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
ENST00000265148.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
ENST00000380026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
ENST00000611174.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
NM_001286734.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
NM_001813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
ENST00000265148.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
ENST00000380026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
ENST00000611174.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
NM_001286734.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
NM_001813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178747.hg.1 |
TC0400011477.hg.1 |
Clariom D TC |