Assay Details
| Gene Symbol: MED14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544000.2 | XP_011542302.1 | 2 - 3 | 158 | 66 | |
| GenBank mRNA | AK308871.1 | - | 2 - 3 | 187 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011544000.2 transcripts.
Set Membership: |
|

MED14
Genomic Map