Assay Details
| Gene Symbol: MUC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024690.2 | NP_078966.2 | 10 - 11 | 36603 | 82 | |
| XM_017027486.1 | XP_016882975.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027487.1 | XP_016882976.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027488.1 | XP_016882977.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027489.1 | XP_016882978.1 | 11 - 12 | 36465 | 82 | ||
| XM_017027490.1 | XP_016882979.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027491.1 | XP_016882980.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027492.1 | XP_016882981.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027493.1 | XP_016882982.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027494.1 | XP_016882983.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027495.1 | XP_016882984.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027496.1 | XP_016882985.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027497.1 | XP_016882986.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027498.1 | XP_016882987.1 | 14 - 15 | 36619 | 82 | ||
| XM_017027499.1 | XP_016882988.1 | 14 - 15 | 27406 | 82 | ||
| XM_017027500.1 | XP_016882989.1 | 13 - 14 | 27325 | 82 | ||
| GenBank mRNA | AF361486.3 | - | 11517 | 82 | ||
| AF414442.2 | - | 36609 | 82 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001139.hg.1
Set Membership: |
|

MUC16
Genomic Map