Assay Details
| Gene Symbol: MUC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF361486.3 | - | 16016 | 97 | ||
| AF414442.2 | - | 45788 | 97 | |||
| AK128681.1 | - | 71 | 97 | |||
| AK128857.1 | - | 518 | 97 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK128681.1 transcripts.
Set Membership: |
|

MUC16
Genomic Map