Assay Details
| Gene Symbol: UBE2U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152489.1 | NP_689702.1 | 2 - 3 | 388 | 151 | |
| XM_005270498.4 | XP_005270555.1 | 2 - 3 | 1182 | 151 | ||
| XM_006710379.3 | XP_006710442.1 | 2 - 3 | 323 | 151 | ||
| XM_011540764.2 | XP_011539066.1 | 3 - 4 | 1123 | 151 | ||
| GenBank mRNA | BC029895.1 | - | 2 - 3 | 388 | 5168406 | 151 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000724.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011503.hg.1 | ENST00000371076.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011503.hg.1 | ENST00000371077.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011503.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011503.hg.1 | TC01000724.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100161011.hg.1 | TC0100008599.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371076.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000371077.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000464349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000608956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | ENST00000611228.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008599.hg.1 | NM_152489.2 | Refseq |

UBE2U
Genomic Map