Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017025155.1 | XP_016880644.1 | 7 - 8 | 802 | 88 | |
| XM_017025156.1 | XP_016880645.1 | 9 - 10 | 1041 | 88 | ||
| GenBank mRNA | AK303282.1 | - | 8 - 9 | 811 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_017025155.1 transcripts.
This assay is in the 5'UTR region of the XM_017025155.1 transcripts.
Set Membership: |
|

DBF4B
Genomic Map