Assay Details
| Gene Symbol: FSTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015082.1 | NP_055897.1 | 6 - 7 | 942 | 53 | |
| XM_011543283.1 | XP_011541585.1 | 6 - 7 | 849 | 53 | ||
| XM_011543284.1 | XP_011541586.1 | 6 - 7 | 920 | 53 | ||
| XM_011543286.2 | XP_011541588.1 | 3 - 4 | 403 | 53 | ||
| GenBank mRNA | AB028984.1 | - | 2 - 3 | 279 | 53 | |
| AK055684.1 | - | 6 - 7 | 920 | 53 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001780.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024198.hg.1 | ENST00000265342.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024198.hg.1 | ENST00000360575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024198.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024198.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024198.hg.1 | TC05001780.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183278.hg.1 | TC0500012044.hg.1 | Clariom D TC |

FSTL4
Genomic Map