Assay Details
| Gene Symbol: CDKN2AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317343.1 | NP_001304272.1 | 1 - 2 | 475 | 81 | |
| NM_017632.3 | NP_060102.1 | 1 - 2 | 475 | 81 | ||
| GenBank mRNA | AF246705.1 | - | 1 - 2 | 423 | 81 | |
| AK000043.1 | - | 1 - 2 | 430 | 81 | ||
| AK096180.1 | - | 1 - 2 | 342 | 81 | ||
| AK225848.1 | - | 1 - 2 | 288 | 81 | ||
| BC022270.1 | - | 1 - 2 | 312 | 4697255 | 81 | |
| CN427056.1 | - | 1 - 2 | 476 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000889.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK096180.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04012131.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012131.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012131.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012131.hg.1 | TC04000889.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000502924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000502924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000502924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012127.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012127.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012127.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012127.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012127.hg.1 | TC04000889.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012130.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012130.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012130.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012130.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012130.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012130.hg.1 | TC04000889.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012128.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012128.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012128.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012128.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012128.hg.1 | TC04000889.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012126.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012126.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012126.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012126.hg.1 | TC04000889.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164788.hg.1 | TC0400009458.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164786.hg.1 | TC0400009458.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164785.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164785.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164785.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164785.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164785.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164785.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164785.hg.1 | TC0400009458.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000502924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000502924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000502924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009458.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164787.hg.1 | TC0400009458.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000302350.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000506835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164790.hg.1 | TC0400009458.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164791.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164791.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164791.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164791.hg.1 | ENST00000504169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164791.hg.1 | ENST00000510928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164791.hg.1 | NM_017632.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164791.hg.1 | TC0400009458.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 168565 | - | - |

CDKN2AIP
Genomic Map