| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021486.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021486.hg.1 |
ENST00000540138.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021486.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021486.hg.1 |
TC06001782.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011996.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182515.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182515.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182515.hg.1 |
ENST00000274813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182515.hg.1 |
NM_000255.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182515.hg.1 |
TC0600011996.hg.1 |
Clariom D TC |