Assay Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030912.2 | NP_112174.2 | 2 - 3 | 786 | 65 | |
| GenBank mRNA | AF220034.1 | - | 2 - 3 | 663 | 65 | |
| AF281046.1 | - | 2 - 3 | 672 | 65 | ||
| AL360160.1 | - | 2 - 3 | 672 | 65 | ||
| BC021925.1 | - | 2 - 3 | 785 | 4857393 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000762.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of autophagy
protein ubiquitination
stem cell population maintenance
negative regulation of viral transcription
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
negative regulation of viral entry into host cell
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
interferon-gamma-mediated signaling pathway
positive regulation of protein localization to nucleus
negative regulation of viral release from host cell
zinc ion binding
ligase activity
identical protein binding
protein homodimerization activity
positive regulation of autophagy
protein ubiquitination
stem cell population maintenance
negative regulation of viral transcription
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
negative regulation of viral entry into host cell
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
interferon-gamma-mediated signaling pathway
positive regulation of protein localization to nucleus
negative regulation of viral release from host cell
zinc ion binding
ligase activity
identical protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009050.hg.1 | ENST00000302424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009050.hg.1 | ENST00000369896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009050.hg.1 | ENST00000462202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009050.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009050.hg.1 | TC10000762.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000479004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008744.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | ENST00000302424.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | ENST00000462202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | ENST00000487927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | NM_030912.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159839.hg.1 | TC1000008744.hg.1 | Clariom D TC |

TRIM8
Genomic Map