Assay Details
| Gene Symbol: SCARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204255.1 | NP_001191184.1 | 1 - 2 | 466 | 67 | |
| NM_005506.3 | NP_005497.1 | 1 - 2 | 466 | 67 | ||
| GenBank mRNA | AK296519.1 | - | 1 - 2 | 170 | 67 | |
| AK313016.1 | - | 1 - 2 | 176 | 67 | ||
| BC021892.1 | - | 1 - 2 | 132 | 3872778 | 67 | |
| BQ924546.1 | - | 1 - 2 | 449 | 67 | ||
| BT006939.1 | - | 1 - 2 | 110 | 67 | ||
| D12676.1 | - | 1 - 2 | 361 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001308.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04017975.hg.1 | ENST00000264896.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017975.hg.1 | ENST00000452464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017975.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017975.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017975.hg.1 | TC04001308.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017978.hg.1 | ENST00000264896.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017978.hg.1 | ENST00000452464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017978.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017978.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017978.hg.1 | TC04001308.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017980.hg.1 | ENST00000264896.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017980.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017980.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017980.hg.1 | TC04001308.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017977.hg.1 | ENST00000264896.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017977.hg.1 | ENST00000452464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017977.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017977.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017977.hg.1 | TC04001308.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175410.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175410.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175410.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175410.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175410.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175410.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175410.hg.1 | TC0400011057.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175406.hg.1 | TC0400011057.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175404.hg.1 | TC0400011057.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175408.hg.1 | TC0400011057.hg.1 | Clariom D TC |

SCARB2
Genomic Map