Assay Details
| Gene Symbol: SCARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005506.3 | NP_005497.1 | 3 - 4 | 783 | 106 | |
| GenBank mRNA | AK313016.1 | - | 3 - 4 | 493 | 106 | |
| BC021892.1 | - | 3 - 4 | 449 | 3872778 | 106 | |
| BT006939.1 | - | 3 - 4 | 427 | 106 | ||
| D12676.1 | - | 3 - 4 | 678 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001308.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04017972.hg.1 | ENST00000264896.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017972.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017972.hg.1 | TC04001308.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017970.hg.1 | ENST00000264896.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017970.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017970.hg.1 | TC04001308.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400175399.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175399.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175399.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175399.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175399.hg.1 | TC0400011057.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400175401.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175401.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175401.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175401.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175401.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175401.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175401.hg.1 | TC0400011057.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000264896.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000452464.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000502908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000509994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | ENST00000511129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_001204255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011057.hg.1 | NM_005506.3 | Refseq |

SCARB2
Genomic Map