Assay Details
| Gene Symbol: HLA-DPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242524.1 | NP_001229453.1 | 5 - 6 | 995 | 114 | |
| NM_001242525.1 | NP_001229454.1 | 5 - 6 | 919 | 114 | ||
| NM_033554.3 | NP_291032.2 | 4 - 5 | 904 | 114 | ||
| GenBank mRNA | BC009956.2 | - | 4 - 5 | 828 | 4309471 | 114 |
| X00457.1 | - | 4 - 5 | 717 | 114 | ||
More Information
Set Membership: |
|

HLA-DPA1
Genomic Map