Assay Details
| Gene Symbol: ENKUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270383.1 | NP_001257312.1 | 3 - 4 | 582 | 94 | |
| NM_145010.3 | NP_659447.1 | 3 - 4 | 669 | 94 | ||
| NR_072992.1 | 3 - 4 | 669 | 94 | |||
| XM_006717389.1 | XP_006717452.1 | 3 - 4 | 665 | 94 | ||
| XM_006717390.1 | XP_006717453.1 | 2 - 3 | 488 | 94 | ||
| XM_011519348.1 | XP_011517650.1 | 3 - 4 | 665 | 94 | ||
| XM_017015830.1 | XP_016871319.1 | 2 - 3 | 488 | 94 | ||
| GenBank mRNA | AK095021.1 | - | 3 - 4 | 614 | 94 | |
| AK292495.1 | - | 3 - 4 | 666 | 94 | ||
| AY454125.1 | - | 3 - 4 | 656 | 94 | ||
| BC026165.2 | - | 3 - 4 | 671 | 4837530 | 94 | |
| CR936720.1 | - | 3 - 4 | 582 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001111.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013975.hg.1 | ENST00000331161.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013975.hg.1 | ENST00000376363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013975.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013975.hg.1 | TC10001111.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168638.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168638.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168638.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168638.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168638.hg.1 | TC1000010071.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168637.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168637.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168637.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168637.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168637.hg.1 | TC1000010071.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168629.hg.1 | TC1000010071.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168636.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168636.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168636.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168636.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168636.hg.1 | TC1000010071.hg.1 | Clariom D TC |

ENKUR
Genomic Map