| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003752.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003752.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003752.hg.1 |
TC16000253.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007215.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151900.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151900.hg.1 |
NM_001039.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151900.hg.1 |
TC1600007215.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600151900.hg.1 |
ENST00000300061.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151900.hg.1 |
NM_001039.3 |
Refseq |