| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038861.hg.1 |
ENST00000283243.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038861.hg.1 |
ENST00000460710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038861.hg.1 |
NM_001195641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038861.hg.1 |
NM_007366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038861.hg.1 |
TC02002468.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038862.hg.1 |
ENST00000283243.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038862.hg.1 |
NM_001195641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038862.hg.1 |
NM_007366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038862.hg.1 |
TC02002468.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200227898.hg.1 |
ENST00000283243.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227898.hg.1 |
NM_001195641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200227898.hg.1 |
NM_007366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200227898.hg.1 |
ENST00000283243.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227898.hg.1 |
NM_001195641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200227898.hg.1 |
NM_007366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200227898.hg.1 |
TC0200016742.hg.1 |
Clariom D TC |