Assay Details
| Gene Symbol: PPP1R10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002714.3 | NP_002705.2 | 13 - 14 | 1876 | 91 | |
| NR_072994.1 | 13 - 14 | 1815 | 91 | |||
| XM_006715130.2 | XP_006715193.1 | 13 - 14 | 1842 | 91 | ||
| XM_011514722.1 | XP_011513024.1 | 14 - 15 | 1866 | 91 | ||
| XM_017010995.1 | XP_016866484.1 | 12 - 13 | 2011 | 91 | ||
| GenBank mRNA | AJ544537.1 | - | 13 - 14 | 1809 | 91 | |
| BC136294.1 | - | 12 - 13 | 1460 | 9020281 | 91 | |
| BC150310.1 | - | 13 - 14 | 1816 | 91 | ||
| Y13247.1 | - | 12 - 13 | 1799 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001502.hg.1, TC6_cox_hap2000137.hg.1, TC6_dbb_hap3000126.hg.1, TC6_mann_hap4000115.hg.1, TC6_mcf_hap5000116.hg.1, TC6_qbl_hap6000127.hg.1, TC6_ssto_hap7000118.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002349.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002349.hg.1 | TC6_qbl_hap6000127.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002350.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002350.hg.1 | TC6_qbl_hap6000127.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016935.hg.1 | ENST00000376511.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016935.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016935.hg.1 | TC06001502.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002376.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002376.hg.1 | TC6_cox_hap2000137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001897.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001897.hg.1 | TC6_mann_hap4000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002067.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002067.hg.1 | TC6_mcf_hap5000116.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002151.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002151.hg.1 | TC6_ssto_hap7000118.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002066.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002066.hg.1 | TC6_mcf_hap5000116.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016936.hg.1 | ENST00000376511.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016936.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016936.hg.1 | TC06001502.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002307.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002307.hg.1 | TC6_dbb_hap3000126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001896.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001896.hg.1 | TC6_mann_hap4000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002377.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002377.hg.1 | TC6_cox_hap2000137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002306.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002306.hg.1 | TC6_dbb_hap3000126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002150.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002150.hg.1 | TC6_ssto_hap7000118.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176623.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176623.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176623.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176623.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176623.hg.1 | TC0600011372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176624.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176624.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176624.hg.1 | TC0600011372.hg.1 | Clariom D TC |

PPP1R10
Genomic Map