Assay Details
| Gene Symbol: PPP1R10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002714.3 | NP_002705.2 | 1 - 2 | 82 | 62 | |
| NR_072994.1 | 1 - 2 | 82 | 62 | |||
| GenBank mRNA | BC150310.1 | - | 1 - 2 | 82 | 62 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_002714.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001920.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001920.hg.1 | TC6_mann_hap4000115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002090.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002090.hg.1 | TC6_mcf_hap5000116.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002174.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002174.hg.1 | TC6_ssto_hap7000118.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002330.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002330.hg.1 | TC6_dbb_hap3000126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002400.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002400.hg.1 | TC6_cox_hap2000137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002373.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002373.hg.1 | TC6_qbl_hap6000127.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016961.hg.1 | ENST00000376511.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016961.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016961.hg.1 | ENST00000537132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016961.hg.1 | NM_002714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016961.hg.1 | TC06001502.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000376511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000461593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000468181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000473954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000476704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000484449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011372.hg.1 | ENST00000496955.1 | Ensembl |

PPP1R10
Genomic Map