| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000461593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000468181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000473954.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000476704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000484449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011372.hg.1 |
ENST00000496955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600176644.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600176644.hg.1 |
ENST00000376511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600176644.hg.1 |
TC0600011372.hg.1 |
Clariom D TC |