| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008579.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008579.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008579.hg.1 |
TC05000615.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500158895.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158895.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158895.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500158895.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158895.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158895.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500158895.hg.1 |
TC0500008582.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000173527.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
ENST00000514194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008582.hg.1 |
NM_016048.2 |
Refseq |