Assay Details
| Gene Symbol: CUBN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081.3 | NP_001072.2 | 36 - 37 | 5391 | 97 | |
| XM_011519708.2 | XP_011518010.1 | 36 - 37 | 6066 | 97 | ||
| XM_011519709.2 | XP_011518011.1 | 10 - 11 | 1754 | 97 | ||
| XM_011519710.2 | XP_011518012.1 | 10 - 11 | 1397 | 97 | ||
| XM_011519711.2 | XP_011518013.1 | 9 - 10 | 2078 | 97 | ||
| GenBank mRNA | AB209880.1 | - | 9 - 10 | 1458 | 97 | |
| AF034611.1 | - | 36 - 37 | 5365 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001070.hg.1
Set Membership: |
|

CUBN
Genomic Map