Assay Details
| Gene Symbol: CUBN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081.3 | NP_001072.2 | 38 - 39 | 5790 | 104 | |
| XM_011519708.2 | XP_011518010.1 | 38 - 39 | 6465 | 104 | ||
| XM_011519709.2 | XP_011518011.1 | 12 - 13 | 2153 | 104 | ||
| XM_011519710.2 | XP_011518012.1 | 12 - 13 | 1796 | 104 | ||
| XM_011519711.2 | XP_011518013.1 | 11 - 12 | 2477 | 104 | ||
| GenBank mRNA | AB209880.1 | - | 11 - 12 | 1857 | 104 | |
| AF034611.1 | - | 38 - 39 | 5764 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001070.hg.1
Set Membership: |
|

CUBN
Genomic Map