Assay Details
| Gene Symbol: MORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014429.3 | NP_055244.3 | 5 - 6 | 403 | 90 | |
| XM_005247362.2 | XP_005247419.1 | 5 - 6 | 360 | 90 | ||
| XM_011512691.1 | XP_011510993.1 | 5 - 6 | 360 | 90 | ||
| XM_011512692.1 | XP_011510994.1 | 5 - 6 | 360 | 90 | ||
| XM_011512693.1 | XP_011510995.1 | 5 - 6 | 360 | 90 | ||
| XM_017006169.1 | XP_016861658.1 | 5 - 6 | 360 | 90 | ||
| GenBank mRNA | AF084946.1 | - | 5 - 6 | 378 | 90 | |
| AK302642.1 | - | 5 - 6 | 360 | 90 | ||
| BC028680.1 | - | 5 - 6 | 404 | 90 | ||
| BC050307.1 | - | 5 - 6 | 342 | 4838254 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001635.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029561.hg.1 | ENST00000232603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029561.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029561.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029561.hg.1 | TC03001635.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189011.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189011.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189011.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189011.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189011.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189011.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189011.hg.1 | TC0300011951.hg.1 | Clariom D TC |

MORC1
Genomic Map