Assay Details
| Gene Symbol: MORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014429.3 | NP_055244.3 | 9 - 10 | 898 | 150 | |
| XM_005247362.2 | XP_005247419.1 | 9 - 10 | 855 | 150 | ||
| XM_011512691.1 | XP_011510993.1 | 9 - 10 | 861 | 150 | ||
| XM_011512692.1 | XP_011510994.1 | 9 - 10 | 861 | 150 | ||
| XM_011512693.1 | XP_011510995.1 | 9 - 10 | 861 | 150 | ||
| XM_011512694.1 | XP_011510996.1 | 6 - 7 | 612 | 150 | ||
| XM_011512696.2 | XP_011510998.1 | 3 - 4 | 312 | 150 | ||
| XM_011512697.2 | XP_011510999.1 | 2 - 3 | 480 | 150 | ||
| XM_017006169.1 | XP_016861658.1 | 9 - 10 | 861 | 150 | ||
| XM_017006170.1 | XP_016861659.1 | 2 - 3 | 280 | 150 | ||
| GenBank mRNA | AF084946.1 | - | 9 - 10 | 873 | 150 | |
| AK302642.1 | - | 9 - 10 | 855 | 150 | ||
| AL353955.1 | - | 3 - 4 | 258 | 150 | ||
| BC028680.1 | - | 9 - 10 | 899 | 150 | ||
| BC050307.1 | - | 9 - 10 | 837 | 4838254 | 150 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001635.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011512697.2,XM_017006170.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03029557.hg.1 | ENST00000232603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029557.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029557.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029557.hg.1 | TC03001635.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011951.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189007.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189007.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189007.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189007.hg.1 | ENST00000232603.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189007.hg.1 | ENST00000483760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189007.hg.1 | NM_014429.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189007.hg.1 | TC0300011951.hg.1 | Clariom D TC |

MORC1
Genomic Map