Assay Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028131.1 | 2 - 3 | 337 | 161 | ||
| GenBank mRNA | BC025683.1 | - | 2 - 3 | 337 | 161 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028131.1 | 2 - 3 | 337 | 161 | ||
| GenBank mRNA | BC025683.1 | - | 2 - 3 | 337 | 161 | |
Set Membership: |
|