Assay Details
| Gene Symbol: SNTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021021.3 | NP_066301.1 | 4 - 5 | 1360 | 59 | |
| GenBank mRNA | AK026095.1 | - | 6 - 7 | 1503 | 59 | |
| AK292655.1 | - | 4 - 5 | 1434 | 59 | ||
| BC098573.1 | - | 4 - 5 | 1320 | 30925462 | 59 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001565.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08020610.hg.1 | ENST00000395601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020610.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020610.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020610.hg.1 | TC08001565.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179017.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179017.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179017.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179017.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179017.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179017.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179017.hg.1 | TC0800011634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |

SNTB1
Genomic Map