Assay Details
| Gene Symbol: SNTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021021.3 | NP_066301.1 | 6 - 7 | 1755 | 118 | |
| GenBank mRNA | AK026095.1 | - | 8 - 9 | 1898 | 118 | |
| AK292655.1 | - | 6 - 7 | 1829 | 118 | ||
| BC098573.1 | - | 6 - 7 | 1715 | 30925462 | 118 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001565.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012350.hg.1 | ENST00000519841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012350.hg.1 | ENST00000519841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012350.hg.1 | ENST00000519841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020607.hg.1 | ENST00000395601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020607.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020607.hg.1 | TC08001565.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020608.hg.1 | ENST00000395601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020608.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020608.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020608.hg.1 | TC08001565.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020606.hg.1 | ENST00000395601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020606.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020606.hg.1 | TC08001565.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0800012350.hg.1 | ENST00000519841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012350.hg.1 | ENST00000519841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012350.hg.1 | ENST00000519841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179012.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179012.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179012.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179012.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179012.hg.1 | TC0800011634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800179014.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179014.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179014.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179014.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179014.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179014.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179014.hg.1 | TC0800011634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800179015.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179015.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179015.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179015.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179015.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179015.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179015.hg.1 | TC0800011634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800179013.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179013.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179013.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179013.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179013.hg.1 | TC0800011634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000395601.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000517992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000519298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | ENST00000520717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011634.hg.1 | NM_021021.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 107347 | - | - |

SNTB1
Genomic Map