| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000310828.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000360141.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000392625.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000577511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000582319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
NM_001256841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
NM_007261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
TC1700008852.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000310828.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000360141.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000392625.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000577511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
ENST00000582319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
NM_001256841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166125.hg.1 |
NM_007261.3 |
Refseq |