Assay Details
| Gene Symbol: GPSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321038.1 | NP_001307967.1 | 7 - 8 | 1053 | 113 | |
| NM_001321039.1 | NP_001307968.1 | 7 - 8 | 1285 | 113 | ||
| NM_013296.4 | NP_037428.3 | 7 - 8 | 1285 | 113 | ||
| XM_006710589.2 | XP_006710652.1 | 6 - 7 | 1744 | 113 | ||
| XM_011541301.2 | XP_011539603.1 | 8 - 9 | 1562 | 113 | ||
| XM_011541302.2 | XP_011539604.1 | 8 - 9 | 2224 | 113 | ||
| XM_011541303.2 | XP_011539605.1 | 7 - 8 | 1980 | 113 | ||
| XM_017001097.1 | XP_016856586.1 | 8 - 9 | 2130 | 113 | ||
| XM_017001098.1 | XP_016856587.1 | 8 - 9 | 1898 | 113 | ||
| GenBank mRNA | AB445462.1 | - | 7 - 8 | 1285 | 113 | |
| AK295563.1 | - | 7 - 8 | 872 | 113 | ||
| AY136740.1 | - | 6 - 7 | 768 | 113 | ||
| BC027732.2 | - | 7 - 8 | 1061 | 5020191 | 113 | |
| CR456786.1 | - | 6 - 7 | 768 | 113 | ||
| KX085485.1 | - | 7 - 8 | 1285 | 113 | ||
| U54999.1 | - | 6 - 7 | 941 | 113 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000936.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
mitotic spindle organization
G-protein coupled receptor signaling pathway
regulation of catalytic activity
maintenance of centrosome location
lung epithelial cell differentiation
nucleotide binding
G-protein alpha-subunit binding
GDP-dissociation inhibitor activity
protein binding
identical protein binding
establishment of mitotic spindle orientation
mitotic spindle organization
G-protein coupled receptor signaling pathway
regulation of catalytic activity
maintenance of centrosome location
lung epithelial cell differentiation
nucleotide binding
G-protein alpha-subunit binding
GDP-dissociation inhibitor activity
protein binding
identical protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014721.hg.1 | ENST00000264126.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014721.hg.1 | ENST00000406462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014721.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014721.hg.1 | TC01000936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000435987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000441735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | ENST00000446797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009333.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166522.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166522.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166522.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166522.hg.1 | ENST00000264126.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166522.hg.1 | ENST00000406462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166522.hg.1 | NM_013296.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166522.hg.1 | TC0100009333.hg.1 | Clariom D TC |

GPSM2
Genomic Map