Assay Details
| Gene Symbol: MRPL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004891.3 | NP_004882.1 | 2 - 3 | 107 | 68 | |
| GenBank mRNA | AF047440.1 | - | 2 - 3 | 81 | 68 | |
| BC009475.1 | - | 2 - 3 | 59 | 4049053 | 68 | |
| BC020834.1 | - | 2 - 3 | 94 | 4775045 | 68 | |
| BI601959.1 | - | 2 - 3 | 110 | 68 | ||
| CR456888.1 | - | 2 - 3 | 46 | 68 | ||
| CR542117.1 | - | 2 - 3 | 46 | 68 | ||
More Information
Set Membership: |
|

MRPL33
Genomic Map