| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012319.hg.1 |
ENST00000379548.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012319.hg.1 |
ENST00000379565.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012319.hg.1 |
ENST00000540702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012319.hg.1 |
ENST00000544447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012319.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012319.hg.1 |
TC0X000911.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012318.hg.1 |
ENST00000379548.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012318.hg.1 |
ENST00000379565.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012318.hg.1 |
ENST00000540702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012318.hg.1 |
ENST00000544447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012318.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012318.hg.1 |
TC0X000911.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00163374.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163374.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163374.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163374.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163374.hg.1 |
TC0X00009218.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00163375.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163375.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163375.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163375.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163375.hg.1 |
TC0X00009218.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000379565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000438357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000457145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000474266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000479809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
ENST00000492128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009218.hg.1 |
NM_004586.2 |
Refseq |