Assay Details
| Gene Symbol: NUP133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018230.2 | NP_060700.2 | 21 - 22 | 3071 | 98 | |
| GenBank mRNA | AF052123.1 | - | 4 - 5 | 674 | 98 | |
| AK001676.1 | - | 21 - 22 | 3071 | 98 | ||
| AK001754.1 | - | 13 - 14 | 1909 | 98 | ||
| AK022572.1 | - | 9 - 10 | 1317 | 98 | ||
| AK092339.1 | - | 20 - 21 | 2815 | 98 | ||
| AK304061.1 | - | 22 - 23 | 3038 | 98 | ||
| AK304861.1 | - | 22 - 23 | 2972 | 98 | ||
| AK314431.1 | - | 21 - 22 | 3028 | 98 | ||
| BC020107.1 | - | 21 - 22 | 3055 | 3919973 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003947.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription-dependent tethering of RNA polymerase II gene DNA at nuclear periphery
chromatin organization
regulation of transcription, DNA-templated
mRNA export from nucleus
tRNA export from nucleus
protein import into nucleus
nuclear pore organization
sister chromatid cohesion
mitotic nuclear envelope disassembly
regulation of glucose transport
viral process
protein sumoylation
viral transcription
neural tube development
neurogenesis
gene silencing by RNA
nuclear pore distribution
paraxial mesoderm development
somite development
intracellular transport of virus
regulation of cellular response to heat
nucleocytoplasmic transporter activity
protein binding
structural constituent of nuclear pore
transcription-dependent tethering of RNA polymerase II gene DNA at nuclear periphery
chromatin organization
regulation of transcription, DNA-templated
mRNA export from nucleus
tRNA export from nucleus
protein import into nucleus
nuclear pore organization
sister chromatid cohesion
mitotic nuclear envelope disassembly
regulation of glucose transport
viral process
protein sumoylation
viral transcription
neural tube development
neurogenesis
gene silencing by RNA
nuclear pore distribution
paraxial mesoderm development
somite development
intracellular transport of virus
regulation of cellular response to heat
nucleocytoplasmic transporter activity
protein binding
structural constituent of nuclear pore
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01059843.hg.1 | ENST00000261396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059843.hg.1 | ENST00000366679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059843.hg.1 | ENST00000366681.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059843.hg.1 | ENST00000537506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059843.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059843.hg.1 | TC01003947.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000434311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000434311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000366678.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000434311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000434311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000485119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | ENST00000490352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017664.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233361.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233361.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233361.hg.1 | ENST00000261396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233361.hg.1 | NM_018230.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233361.hg.1 | TC0100017664.hg.1 | Clariom D TC |

NUP133
Genomic Map