| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
ENST00000265755.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
ENST00000424337.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
ENST00000455841.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
ENST00000476977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
ENST00000489094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
NM_001199207.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
NM_005685.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
NM_016328.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006213.hg.1 |
TC07000455.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000265755.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000424337.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000455841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000476977.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000489094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
NM_001199207.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
NM_005685.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
NM_016328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000265755.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000424337.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000455841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000476977.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
ENST00000489094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
NM_001199207.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
NM_005685.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
NM_016328.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700155357.hg.1 |
TC0700008004.hg.1 |
Clariom D TC |