Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae) pseudogene |
LOC100303749 | -23.3kb | Anti-sense | Chr. 5 - 65944805 - 65945573 on NCBI Build GRCh38 | |
| Downstream | splicing regulatory glutamic acid and lysine rich protein 1 | SREK1 |
SFRS12, SRrp508, SRrp86 |
171.8kb | Sense | Chr. 5 - 66144218 - 66183616 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map