Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5787 | MIR5787 | hsa-mir-5787 | -12.1kb | Anti-sense | Chr. 3 - 50227436 - 50227490 on NCBI Build GRCh38 |
| Downstream | SEMA3B antisense RNA 1 (head to head) | SEMA3B-AS1 | 26.7kb | Sense | Chr. 3 - 50266641 - 50267371 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map