| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020009.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020009.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020009.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020009.hg.1 |
TC06001679.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011674.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180114.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180114.hg.1 |
ENST00000483552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180114.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180114.hg.1 |
ENST00000373699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180114.hg.1 |
TC0600011674.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600180114.hg.1 |
NM_016059.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180114.hg.1 |
ENST00000483552.1 |
Ensembl |