Assay Details
| Gene Symbol: TMPRSS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002772.2 | NP_002763.2 | 3 - 4 | 378 | 154 | |
| XM_011529659.1 | XP_011527961.1 | 3 - 4 | 497 | 154 | ||
| GenBank mRNA | BC111749.1 | - | 3 - 4 | 367 | 40008569 | 154 |
| DB242953.1 | - | 3 - 4 | 497 | 154 | ||
| U09860.1 | - | 3 - 4 | 387 | 154 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000305.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21003659.hg.1 | ENST00000284885.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003659.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003659.hg.1 | TC21000305.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003661.hg.1 | ENST00000284885.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003661.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003661.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003661.hg.1 | TC21000305.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | TC2100007676.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152052.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152050.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152050.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152050.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152050.hg.1 | TC2100007676.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152050.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152050.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152050.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |

TMPRSS15
Genomic Map