Assay Details
| Gene Symbol: DCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006719270.1 | XP_006719333.1 | 1 - 2 | 48 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006719270.1 transcripts.
Set Membership: |
|

DCN
Genomic Map