Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lysine methyltransferase 5A | KMT5A |
PR-Set7, SET07, SET8, SETD8 |
Anti-sense | Chr. 12 - 123383778 - 123409357 on NCBI Build GRCh38 | |
| Downstream | Rab interacting lysosomal protein like 2 | RILPL2 | RLP2 | 6.4kb | Sense | Chr. 12 - 123408567 - 123436733 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map