Assay Details
| Gene Symbol: ENKUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270383.1 | NP_001257312.1 | 5 - 6 | 895 | 116 | |
| NM_145010.3 | NP_659447.1 | 5 - 6 | 982 | 116 | ||
| NR_072992.1 | 5 - 6 | 982 | 116 | |||
| XM_017015830.1 | XP_016871319.1 | 4 - 5 | 801 | 116 | ||
| GenBank mRNA | AK095021.1 | - | 5 - 6 | 927 | 116 | |
| AK292495.1 | - | 5 - 6 | 979 | 116 | ||
| AY454125.1 | - | 5 - 6 | 969 | 116 | ||
| BC026165.2 | - | 5 - 6 | 984 | 4837530 | 116 | |
| CR936720.1 | - | 5 - 6 | 895 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10013970.hg.1 | ENST00000331161.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013970.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013970.hg.1 | TC10001111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013971.hg.1 | ENST00000331161.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013971.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013971.hg.1 | TC10001111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013972.hg.1 | ENST00000331161.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013972.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013972.hg.1 | TC10001111.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000331161.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000376363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000483339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000496261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | ENST00000615958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_001270383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NM_145010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010071.hg.1 | NR_072993.1 | Refseq |

ENKUR
Genomic Map