| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008269.hg.1 |
ENST00000300452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008269.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008269.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008269.hg.1 |
TC09000685.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000461102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000608951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
ENST00000609948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_001305942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008858.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159401.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159401.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159401.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159401.hg.1 |
ENST00000300452.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159401.hg.1 |
ENST00000372875.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159401.hg.1 |
NM_016035.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159401.hg.1 |
TC0900008858.hg.1 |
Clariom D TC |