Assay Details
| Gene Symbol: C9orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC021261.1 | - | 1 - 2 | 102 | 4136397 | 75 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC021261.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181612.hg.1 | TC0900011938.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181614.hg.1 | TC0900011938.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181616.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181616.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181616.hg.1 | TC0900011938.hg.1 | Clariom D TC |

C9orf116
Genomic Map