Assay Details
| Gene Symbol: IRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002199.3 | NP_002190.2 | 2 - 3 | 293 | 99 | |
| GenBank mRNA | AA361545.1 | - | 2 - 3 | 229 | 99 | |
| AK312953.1 | - | 2 - 3 | 271 | 99 | ||
| BC015803.2 | - | 1 - 2 | 100 | 3920890 | 99 | |
| BP200998.1 | - | 2 - 3 | 293 | 99 | ||
| BT007264.1 | - | 1 - 2 | 93 | 99 | ||
| CR457077.1 | - | 1 - 2 | 93 | 99 | ||
| X15949.1 | - | 2 - 3 | 191 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001805.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
blood coagulation
cell proliferation
positive regulation of transcription from RNA polymerase II promoter
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
blood coagulation
cell proliferation
positive regulation of transcription from RNA polymerase II promoter
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000504340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000504340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024582.hg.1 | ENST00000393593.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024582.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024582.hg.1 | TC04001805.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186276.hg.1 | TC0400012606.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000393593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000502750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000504340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000504340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000505067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000506230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000507523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000509274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000510814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | ENST00000512020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012606.hg.1 | NM_002199.3 | Refseq |

IRF2
Genomic Map