| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057428.hg.1 |
ENST00000367161.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057428.hg.1 |
ENST00000367162.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057428.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057428.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057428.hg.1 |
TC01003743.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
ENST00000367161.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
ENST00000367162.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
ENST00000615388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
NM_015375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
NM_199462.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229206.hg.1 |
TC0100017060.hg.1 |
Clariom D TC |