Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308088.1 | NP_001295017.1 | 2 - 3 | 189 | 70 | |
| XM_011543454.2 | XP_011541756.1 | 2 - 3 | 436 | 70 | ||
| GenBank mRNA | BC038838.1 | - | 1 - 2 | 48 | 5170587 | 70 |
| BM728431.1 | - | 2 - 3 | 189 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC038838.1,NM_001308088.1,XM_011543454.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157974.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157974.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157974.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157974.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157974.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157974.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157974.hg.1 | TC0500008454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157972.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157972.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157972.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157972.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157972.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157972.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157972.hg.1 | TC0500008454.hg.1 | Clariom D TC |

PRR16
Genomic Map