Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300783.1 | NP_001287712.1 | 1 - 2 | 367 | 74 | |
| GenBank mRNA | BC092474.1 | - | 1 - 2 | 368 | 30528467 | 74 |
More Information
Set Membership: |
|

PRR16
Genomic Map