Assay Details
| Gene Symbol: ARFGEF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020340.4 | NP_065073.3 | 15 - 16 | 2739 | 66 | |
| GenBank mRNA | AB033070.2 | - | 6 - 7 | 1653 | 66 | |
| AB252196.1 | - | 15 - 16 | 2739 | 66 | ||
| AF413080.1 | - | 4 - 5 | 1347 | 66 | ||
| AK124312.1 | - | 11 - 12 | 2177 | 66 | ||
| BC137174.1 | - | 13 - 14 | 2374 | 9021171 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001029.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06012055.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012055.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012055.hg.1 | TC06001029.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165847.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165847.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165847.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165847.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165847.hg.1 | TC0600009606.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |

ARFGEF3
Genomic Map